missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzoxazole 98.0+%, TCI America™

Catalog No. P22645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22645G 5 g
P22641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22645G Supplier TCI America Supplier No. P22645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 833-50-1
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00012183
InChI Key FIISKTXZUZBTRC-UHFFFAOYSA-N
Synonym 2-phenylbenzoxazole, 2-phenylbenzo d oxazole, benzoxazole, 2-phenyl, ccris 7874, phenyl-2 benzoxazole french, 2-phenyl-benzoxazole, phenyl-2 benzoxazole, 2-phenybenzoxazole, 2-phenyl benzoxazole, 2-phenyl-benzooxazole
PubChem CID 70030
IUPAC Name 2-phenyl-1,3-benzoxazole
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3O2

Specifications

Melting Point 103°C
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylbenzoxazole
RTECSNumber DM4865700
TSCA Yes