Learn More
2-Phenyl-2-propanol, 98+%
CAS: 617-94-7 | C9H12O | 136.19 g/mol
Supplier: Thermo Scientific Chemicals A1558022
Description
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.
Solubility
Insoluble in water.
Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.
Specifications
| 2-Phenyl-2-propanol | |
| 29°C to 34°C | |
| 215°C | |
| C9H12O | |
| MFCD00004456 | |
| 1905012 | |
| Insoluble in water. | |
| CC(C)(O)C1=CC=CC=C1 | |
| 136.19 | |
| 136.19 |
| 617-94-7 | |
| 0.973 | |
| 87°C (188°F) | |
| 1.5196 | |
| 100 g | |
| 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol | |
| BDCFWIDZNLCTMF-UHFFFAOYSA-N | |
| 2-phenylpropan-2-ol | |
| 12053 | |
| ≥98% |
Chemical Identifiers
| 617-94-7 | |
| 136.19 | |
| BDCFWIDZNLCTMF-UHFFFAOYSA-N | |
| 12053 | |
| CC(C)(O)C1=CC=CC=C1 |
| C9H12O | |
| MFCD00004456 | |
| 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol | |
| 2-phenylpropan-2-ol |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 210-539-5
RTECSNumber : DO4562000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only