accessibility menu, dialog, popup

UserName

CH3H3COH

CAS RN 617-94-7

IUPAC Name: 2-phenylpropan-2-ol
Synonyms: cumyl alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: BDCFWIDZNLCTMF-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=CC=C1
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)