missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, 98+%

Catalog No. p-7022691
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1558014 25 g
AAA1558022 100 g
AAA1558036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1558014 Supplier Thermo Scientific Chemicals Supplier No. A1558014
Only null left

CAS: 617-94-7 | C9H12O | 136.19 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

Solubility
Insoluble in water.

Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1

Specifications

Melting Point 29°C to 34°C
Density 0.973
Boiling Point 215°C
Flash Point 87°C (188°F)
Refractive Index 1.5196
Quantity 25 g
Beilstein 1905012
Solubility Information Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 2-Phenyl-2-propanol
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-539-5
RTECSNumber DO4562000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only