missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-pentenal, (E)+(Z), 90+%

Catalog No. AAA1363514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363514 25 g
AAA1363506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1363514 Supplier Thermo Scientific Chemicals Supplier No. A1363514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3491-63-2 | C11H12O | 160.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3491-63-2
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00051835
InChI Key YPAJRUMMODCONM-IZZDOVSWSA-N
Synonym 2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
PubChem CID 15310469
IUPAC Name (Z)-2-phenylpent-2-enal
SMILES CCC=C(C=O)C1=CC=CC=C1

Specifications

Density 1.005
Boiling Point 124°C to 126°C (14 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.551
Quantity 25 g
Beilstein 1859032
Sensitivity Air sensitive
Formula Weight 160.22
Percent Purity ≥90%
Chemical Name or Material 2-Phenyl-2-pentenal, (E)+(Z)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only