missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxytetrahydropyran 90.0+%, TCI America™

Catalog No. P08535ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08535ML 5 mL
P085325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08535ML Supplier TCI America Supplier No. P08535ML
Only null left

Chemical Identifiers

CAS 4203-50-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00059801
InChI Key SHRDVLUJLDYXSO-UHFFFAOYNA-N
Synonym Tetrahydro-2-phenoxy-2H-pyran
PubChem CID 107240
IUPAC Name 2-phenoxyoxane
SMILES C1CCC(OC2=CC=CC=C2)OC1

Specifications

Quantity 5 mL
Formula Weight 178.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxytetrahydropyran