missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phénoxybenzaldéhyde 96,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 153°C |
| Quantity | 1 g |
| Formula Weight | 198.22 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-Phenoxybenzaldehyde |
Sécurité et manipulation
| TSCA | No |