missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzaldehyde, 98%, Thermo Scientific™

Catalog No. AAH5044803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5044803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5044803 Supplier Thermo Scientific Chemicals Supplier No. H5044803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Phenoxybenzaldehyde is used as a reagent in the synthesis of chiral 1,3,4-oxadiazol-2-ones as potent and highly selective FAAH inhibitors. Also used as a reagent in the synthesis of [4-(phenoxy)pyridin-3-yl]methylamines; a new class of selective noradrenaline reuptake inhibitors (NRI).

Solubility
Very slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 19434-34-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00800666
InChI Key IMPIIVKYTNMBCD-UHFFFAOYSA-N
Synonym o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384
PubChem CID 88060
IUPAC Name 2-phenoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OC1=CC=CC=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Very slightly soluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 2-Phenoxybenzaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 243-061-0
TSCA No
Recommended Storage Ambient temperatures