missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone 99.0+%, TCI America™

Catalog No. P0060500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0060500ML 500 mL
P006025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0060500ML Supplier TCI America Supplier No. P0060500ML

Chemical Identifiers

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00009400
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Specifications

Melting Point -78°C
Color Colorless
Boiling Point 101°C
Quantity 500 mL
UN Number 1249
Formula Weight 86.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentanone
EINECSNumber (2)-0542
RTECSNumber SA7875000
TSCA Yes