missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 98-100% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019470
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019470 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019470 Supplier Merck Emd Millipore Supplier No. 462115ML
Only null left
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 941°F
CAS 107-87-9
Vapor Pressure 27 mmHg (20°C)
Density 0.809 g/mL (at 25°C)
Flash Point 7°C
Physical Form Neat
Linear Formula CH3 COCH2 CH2 CH3
MDL Number MFCD00009400
Melting Point -78°C
Boiling Point 101°C to 105°C
Synonym Methyl propyl ketone
Solubility Soluble in water
Type Analytical Standard
Quantity 5 mL
Formula Weight 86.13
Percent Purity 98 to 100% (GC)
Grade Analytical Standard
Show More Show Less