missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanol 98,0+%, TCI America™

Numéro de catalogue. P005625ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P005625ML 25 mL
P0056500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P005625ML Fournisseur TCI America Code fournisseur P005625ML

Identifiants chimiques

CAS 6032-29-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004579
InChI Key JYVLIDXNZAXMDK-UHFFFAOYSA-N
Synonym 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
PubChem CID 22386
ChEBI CHEBI:77518
IUPAC Name Pentan-2-OL
SMILES CCCC(C)O

Spécifications

Color Incolore
Boiling Point 118°C
Quantity 25 mL
UN Number 1105
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material 2-Pentanol
EINECSNumber (2)-0217
RTECSNumber SA4900000
TSCA Yes