missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)pyridine 97.0+%, TCI America™

Catalog No. T12285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12285G 5 g
T122825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12285G Supplier TCI America Supplier No. T12285G

Chemical Identifiers

CAS 4467-06-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006281
InChI Key KJNZQKYSNAQLEO-UHFFFAOYSA-N
Synonym 2-p-tolyl pyridine, 2-4-methylphenyl pyridine, 2-p-tolylpyridine, 2-4-tolyl pyridine, pyridine, 2-4-methylphenyl, pyridine,2-p-tolyl, 2-p-toluyl pyridine, pubchem16062, 4-2-pyridyl toluene
PubChem CID 78219
IUPAC Name 2-(4-methylphenyl)pyridine
SMILES CC1=CC=C(C=C1)C2=CC=CC=N2

Specifications

Boiling Point 157°C
Quantity 5 g
Formula Weight 169.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(p-Tolyl)pyridine