missing translation for 'onlineSavingsMsg'
Learn More

2-Oxoglutaric Acid 99.0+%, TCI America™

Catalog No. K0005500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K0005500G 500 g
K000525G 25 g
K0005100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K0005500G Supplier TCI America Supplier No. K0005500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-50-7
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00004165
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
PubChem CID 51
ChEBI CHEBI:30915
IUPAC Name 2-oxopentanedioic acid
SMILES C(CC(=O)O)C(=O)C(=O)O

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 500 g
Formula Weight 146.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxoglutaric Acid
EINECSNumber (9)-0467
TSCA Yes