missing translation for 'onlineSavingsMsg'
Learn More

2-Oxocyclohexanepropionitrile, 99%

Numéro de catalogue. AAH5072203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH5072203 1 g
AAH5072206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH5072203 Fournisseur Thermo Scientific Chemicals Code fournisseur H5072203
Il en reste null

CAS: 4594-78-9 | C9H13NO | 151.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 4594-78-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00013797
InChI Key SPTVCXKSSRBTMN-UHFFFAOYNA-N
Synonym 3-2-oxocyclohexyl propanenitrile, 2-2-cyanoethyl cyclohexanone, cyclohexanepropanenitrile, 2-oxo, 2-2-cyanethyl cyclohexanone, 2-oxo-1-cyclohexanepropionitrile, cyclohexanepropionitrile, 2-oxo, 2-oxocyclohexanepropionitrile, 2-.beta.-cyanoethyl cyclohexanone, 2-b-cyanoethyl cyclohexanone, 2-oxocyclohexanepropiononitrile
PubChem CID 96223
IUPAC Name 3-(2-oxocyclohexyl)propanenitrile
SMILES O=C1CCCCC1CCC#N

Spécifications

Density 1.021
Boiling Point 138°C to 142°C (10 mmHg)
Quantity 1 g
UN Number UN3276
Formula Weight 151.21
Percent Purity 99%
Chemical Name or Material 2-Oxocyclohexanepropionitrile
Hazard Category H312+H332
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 224-991-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only