missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-1-imidazolidinecarbonyl Chloride 97.0+%, TCI America™

Catalog No. p-7134291
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03525G Supplier TCI America Supplier No. O03525G

Chemical Identifiers

CAS 13214-53-4
Molecular Formula C4H5ClN2O2
Molecular Weight (g/mol) 148.546
MDL Number MFCD02683529
InChI Key NXJZQSRAFBHNLI-UHFFFAOYSA-N
Synonym 1-Chlorocarbonyl-2-imidazolidinone, 1-Chloroformyl-2-imidazolidinone
PubChem CID 83241
IUPAC Name 2-oxoimidazolidine-1-carbonyl chloride
SMILES C1CN(C(=O)N1)C(=O)Cl

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 5 g
Formula Weight 148.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Oxo-1-imidazolidinecarbonyl Chloride
TSCA No
Recommended Storage Freezer