missing translation for 'onlineSavingsMsg'
Learn More

2-Octene, cis + trans, 98%

Catalog No. AAL0386206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0386206 5 g
AAL0386214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0386206 Supplier Thermo Scientific Chemicals Supplier No. L0386206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-67-1 | C8H16 | 112.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-67-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCCC=CC

Specifications

Density 0.715
Boiling Point 123°C to 125°C
Flash Point 14°C (57°F)
Refractive Index 1.415
Quantity 5 g
UN Number UN3295
Beilstein 1719495
Formula Weight 112.22
Percent Purity 98%
Chemical Name or Material 2-Octene, Cis + Trans
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 203-894-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only