missing translation for 'onlineSavingsMsg'
Learn More

2-Octen-4-one 96.0+%, TCI America™

Catalog No. O03605ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03605ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03605ML Supplier TCI America Supplier No. O03605ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4643-27-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00061023
InChI Key FMDLEUPBHMCPQV-GQCTYLIASA-N
Synonym 2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one
PubChem CID 5365891
IUPAC Name (E)-oct-2-en-4-one
SMILES CCCCC(=O)C=CC

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Octen-4-one
TSCA Yes
Recommended Storage Refrigerator