missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, ≥99.5% (GC), MilliporeSigma™ Supelco™

$244.20 - $244.20

Specifications

CAS 111-13-7
Density 0.819 g/mL (at 25°C)
Flash Point 56°C
Linear Formula CH3(CH2)5 COCH3
MDL Number MFCD00009540
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019133
View Documents
Merck Emd Millipore
024795ML
5 mL
Each for $244.20
Only null left
 
Description

Description

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

111-13-7
56°C
MFCD00009540
173°C
Analytical Standard
≥99.5% (GC)
0.819 g/mL (at 25°C)
CH3(CH2)5 COCH3
-16°C
Hexyl methyl ketone
128.21
Analytical Standard
Videos
SDS
Documents

Documents