missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019133
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019133 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019133 Supplier Merck Emd Millipore Supplier No. 024795ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 111-13-7
Density 0.819 g/mL (at 25°C)
Flash Point 56°C
Linear Formula CH3(CH2)5 COCH3
MDL Number MFCD00009540
Melting Point -16°C
Boiling Point 173°C
Synonym Hexyl methyl ketone
Type Analytical Standard
Quantity 5 mL
Formula Weight 128.21
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less