missing translation for 'onlineSavingsMsg'
Learn More

2'-O-Methylguanosine Hydrate 98.0+%, TCI America™

Catalog No. M2318200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2318200MG 200 mg
M23181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2318200MG Supplier TCI America Supplier No. M2318200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2140-71-8
Molecular Formula C11H15N5O5
Molecular Weight (g/mol) 297.27
MDL Number MFCD00057053
InChI Key OVYNGSFVYRPRCG-YPLCUDRINA-N
Synonym 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
PubChem CID 188959
ChEBI CHEBI:19229
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Specifications

Melting Point 236°C
Color White
Quantity 200 mg
Formula Weight 297.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-O-Methylguanosine Hydrate
TSCA No
Recommended Storage Refrigerator