missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrotoluene 99.0+%, TCI America™

Supplier:  TCI America N027525G

 View more versions of this product

Catalog No. N027525G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Nitrotoluene
Yellow
C7H7NO2
25 g
2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
CC1=CC=CC=C1[N+](=O)[O-]
137.138
CHEBI:33098
≥99.0% (GC)
88-72-2
222°C
MFCD00007157
1664
PLAZTCDQAHEYBI-UHFFFAOYSA-N
1-methyl-2-nitrobenzene
6944
137.14
Liquid

Chemical Identifiers

88-72-2
137.138
PLAZTCDQAHEYBI-UHFFFAOYSA-N
6944
1-methyl-2-nitrobenzene
C7H7NO2
MFCD00007157
2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
CHEBI:33098
CC1=CC=CC=C1[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0437

RTECSNumber : XT3150000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions