missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrotoluene 99.0+%, TCI America™

Catalog No. p-7133721
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N027525G 25 g
N0275500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N027525G Supplier TCI America Supplier No. N027525G
Only null left

Chemical Identifiers

CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007157
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 1664
Formula Weight 137.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitrotoluene
EINECSNumber (3)-0437
RTECSNumber XT3150000
TSCA Yes