accessibility menu, dialog, popup

UserName

CH3NOO

CAS RN 88-72-2

IUPAC Name: 1-methyl-2-nitrobenzene
Synonyms: 2-nitrotoluene ONT
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: PLAZTCDQAHEYBI-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)