missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrothiophene (contains 3-Nitrothiophene) 80.0+%, TCI America™

Catalog No. N04075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04075G 5 g
N040725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04075G Supplier TCI America Supplier No. N04075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-40-5
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.133
MDL Number MFCD00005425
InChI Key JIZRGGUCOQKGQD-UHFFFAOYSA-N
Synonym thiophene, 2-nitro, 2-nitro-thiophene, 2-nitro thiophene, 2-nitrothiophene, tech, nitrothiophene, 2-nitrothiofene, pubchem5570, 2-nitrothiophene, tech., acmc-1aw2t, wln: t5sj bnw
PubChem CID 11866
IUPAC Name 2-nitrothiophene
SMILES C1=CSC(=C1)[N+](=O)[O-]

Specifications

Melting Point 43°C
Boiling Point 224°C
Quantity 5 g
Formula Weight 129.13
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrothiophene (contains 3-Nitrothiophene)
RTECSNumber XN0175000
TSCA Yes