missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitroso-1-naphthol 98.0+%, TCI America™
$119.30 - $385.86
Chemical Identifiers
| CAS | 132-53-6 |
|---|---|
| Molecular Formula | C10H7NO2 |
| Molecular Weight (g/mol) | 173.17 |
| MDL Number | MFCD00003932 |
| InChI Key | SYUYTOYKQOAVDW-UHFFFAOYSA-N |
| Synonym | 2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime |
| PubChem CID | 8592 |
| IUPAC Name | 2-nitrosonaphthalen-1-ol |
| SMILES | OC1=C2C=CC=CC2=CC=C1N=O |
Chemical Identifiers
| 132-53-6 | |
| 173.17 | |
| SYUYTOYKQOAVDW-UHFFFAOYSA-N | |
| 8592 | |
| OC1=C2C=CC=CC2=CC=C1N=O |
| C10H7NO2 | |
| MFCD00003932 | |
| 2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime | |
| 2-nitrosonaphthalen-1-ol |
Specifications
| 132-53-6 | |
| C10H7NO2 | |
| 5 g | |
| SYUYTOYKQOAVDW-UHFFFAOYSA-N | |
| 2-nitrosonaphthalen-1-ol | |
| 8592 | |
| ≥98.0% (GC,T) | |
| 2-Nitroso-1-naphthol |
| 148°C | |
| MFCD00003932 | |
| 2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime | |
| OC1=C2C=CC=CC2=CC=C1N=O | |
| 173.17 | |
| 173.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QL4550000
TSCA : Yes