accessibility menu, dialog, popup

UserName

CAS RN 132-53-6

IUPAC Name: 2-nitrosonaphthalen-1-ol
Synonyms: 2-nitroso-1-naphthol
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: SYUYTOYKQOAVDW-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=CC=C1N=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results