missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroso-1-naphthol 98.0+%, TCI America™

Supplier:  TCI America N026725G

 View more versions of this product

Catalog No. N026725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Nitroso-1-naphthol
148°C
MFCD00003932
2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime
OC1=C2C=CC=CC2=CC=C1N=O
173.17
173.17
Crystalline Powder
132-53-6
C10H7NO2
25 g
SYUYTOYKQOAVDW-UHFFFAOYSA-N
2-nitrosonaphthalen-1-ol
8592
≥98.0% (GC,T)

Chemical Identifiers

132-53-6
173.17
SYUYTOYKQOAVDW-UHFFFAOYSA-N
8592
OC1=C2C=CC=CC2=CC=C1N=O
C10H7NO2
MFCD00003932
2-nitroso-1-naphthol, 2-nitrosonaphthol, 1-naphthalenol, 2-nitroso, 1-naphthol, 2-nitroso, 1,2-naphthoquinone 2-oxime, nitrosonaphthol, .beta.-nitrosonaphthol, unii-cc0s5ho41v, beta-nitroso-alpha-naphthol, 1,2-naphthalenedione, 2-oxime
2-nitrosonaphthalen-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QL4550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions