missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylhydrazine, 97%, moistened with ca 30% water

Catalog No. AC128830100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128830100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC128830100 Supplier Thermo Scientific Chemicals Supplier No. 128830100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3034-19-3 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-19-3 , 7732-18-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007577
InChI Key FRBUNLLUASHNDJ-UHFFFAOYSA-N
Synonym 2-nitrophenyl hydrazine, hydrazine, nitrophenyl, hydrazine, o-nitrophenyl, o-nitrophenylhydrazine, hydrazine, 2-nitrophenyl, 2-nitrophenylhrdrazine, 2-nitrophenyl diazane, pubchem12859, acmc-1agti, o-nitrophenyl hydrazine
PubChem CID 3314739
IUPAC Name (2-nitrophenyl)hydrazine
SMILES C1=CC=C(C(=C1)NN)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 89.0°C to 94.0°C
Color Orange to Red
Loss on Drying 30% to 35%
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4NHNH2
Quantity 10 g
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene
Formula Weight 153.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Nitrophenylhydrazine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 221-222-6

RUO – Research Use Only