accessibility menu, dialog, popup

UserName

ClH3NNHNOO

CAS RN 3034-19-3

IUPAC Name: 2-(2-nitrophenyl)hydrazinium chloride
Synonyms: 2-(2-nitrophenyl)hydrazinium chloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C6H8ClN3O2
InChi Key: XCUBVSAYUSFHNN-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]NC1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results