missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol 93.0+%, TCI America™

Catalog No. N03685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03685G 5 g
N036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03685G Supplier TCI America Supplier No. N03685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-48-9
Molecular Formula C2H5NO3
Molecular Weight (g/mol) 91.066
MDL Number MFCD00007405
InChI Key KIPMDPDAFINLIV-UHFFFAOYSA-N
Synonym ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
PubChem CID 12252
IUPAC Name 2-nitroethanol
SMILES C(CO)[N+](=O)[O-]

Specifications

Boiling Point 197°C
Quantity 5 g
Formula Weight 91.07
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitroethanol
RTECSNumber KL9370000
TSCA No