missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, 97%

Numéro de catalogue. p-3779302
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC397960050 5 g Glass bottle
AC397960250 25 g Glass bottle
AC397961000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC397960050 Fournisseur Thermo Scientific Chemicals Code fournisseur 397960050
Il en reste null

CAS: 625-48-9 | C2H5NO3 | 91.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 625-48-9
Molecular Formula C2H5NO3
Molecular Weight (g/mol) 91.07
MDL Number MFCD00007405
InChI Key KIPMDPDAFINLIV-UHFFFAOYSA-N
Synonym ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
PubChem CID 12252
IUPAC Name 2-nitroethanol
SMILES C(CO)[N+](=O)[O-]

Spécifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Yellow
Density 1.2920g/mL
Boiling Point 203.0°C to 206.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4445
Linear Formula O2NCH2CH2OH
Quantity 5 g
Specific Gravity 1.292
Solubility Information Solubility in water: miscible
Formula Weight 91.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Nitroethanol
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-895-1

RUO – Research Use Only