missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrocinnamaldehyde 98.0+%, TCI America™

Catalog No. N06115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06115G 5 g
N061125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06115G Supplier TCI America Supplier No. N06115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1466-88-2
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007188
InChI Key VMSMELHEXDVEDE-HWKANZROSA-N
Synonym 2-nitrocinnamaldehyde, 3-2-nitrophenyl acrylaldehyde, o-nitrocinnamaldehyde, trans-2-nitrocinnamaldehyde, 2e-3-2-nitrophenyl prop-2-enal, 2-propenal, 3-2-nitrophenyl, e-3-2-nitrophenyl acrylaldehyde, e-3-2-nitrophenyl prop-2-enal, 3-2-nitrophenyl prop-2-enal, ccris 3775
PubChem CID 5367122
IUPAC Name (E)-3-(2-nitrophenyl)prop-2-enal
SMILES C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]

Specifications

Melting Point 128°C
Quantity 5 g
Formula Weight 177.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrocinnamaldehyde