missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl Cyclohexylcarbamate 98.0+%, TCI America™

Catalog No. N10521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10521G Supplier TCI America Supplier No. N10521G

Chemical Identifiers

CAS 119137-03-0
Molecular Formula C14H18N2O4
Molecular Weight (g/mol) 278.308
MDL Number MFCD22376660
InChI Key NJMCHQONLVUNAM-UHFFFAOYSA-N
Synonym Cyclohexylcarbamic Acid 2-Nitrobenzyl Ester
PubChem CID 21910133
IUPAC Name (2-nitrophenyl)methyl N-cyclohexylcarbamate
SMILES C1CCC(CC1)NC(=O)OCC2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 104°C
Color Yellow
Quantity 1 g
Formula Weight 278.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzyl Cyclohexylcarbamate