missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl Bromure 98,0+%, TCI America™

Numéro de catalogue. N04045G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N04045G 5 g
N040425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. N04045G Fournisseur TCI America Code fournisseur N04045G

Identifiants chimiques

CAS 3958-60-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007184
InChI Key HXBMIQJOSHZCFX-UHFFFAOYSA-N
Synonym 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
PubChem CID 77569
IUPAC Name 1-(bromométhyl)-2-nitrobenzène
SMILES C1=CC=C(C(=C1)CBr)[N+](=O)[O-]

Spécifications

Melting Point 46°C
Color Jaune
Quantity 5 g
UN Number 3261
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2-Nitrobenzyl Bromide