missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzotrifluoride, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | 2-Nitrobenzotrifluoride |
| Point d’ébullition | 216.0°C to 217.0°C |
| Point d’éclair | 95°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (GC) |
| Emballage | Glass Bottle |
| Formule linéaire | O2NC6H4CF3 |
| Quantité | 1g |
| Beilstein | 05,II,251 |
| Poids de formule | 191.11 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated cl |
| EINECSNumber | 206-855-8 |
RUO – Research Use Only