missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroanisole, 99%

Catalog No. p-7024586
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1951836 500 g
AAA195180E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1951836 Supplier Thermo Scientific Chemicals Supplier No. A1951836

CAS: 91-23-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 91-23-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007096
InChI Key CFBYEGUGFPZCNF-UHFFFAOYSA-N
Synonym 2-nitroanisole, o-nitroanisole, anisole, o-nitro, 1-nitro-2-methoxybenzene, 2-methoxynitrobenzene, o-nitrophenyl methyl ether, benzene, 1-methoxy-2-nitro, 2-methoxy-1-nitrobenzene, benzene, methoxynitro, o-nitro methoxy benzene
PubChem CID 7048
ChEBI CHEBI:48722
IUPAC Name 1-methoxy-2-nitrobenzene
SMILES COC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 9°C to 10°C
Density 1.254
Boiling Point 272°C to 273°C
Flash Point 142°C (287°F)
Refractive Index 1.561
Quantity 500 g
UN Number UN2730
Beilstein 1868032
Merck Index 14,6585
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 2-Nitroanisole
Show More Show Less
Hazard Category H302-H350
Hazard Statement GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROANISOLE, LIQUID
EINECSNumber 202-052-1
RTECSNumber BZ8790000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only