missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-p-tolunitrile 98.0+%, TCI America™

Catalog No. N04945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04945G 5 g
N04941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04945G Supplier TCI America Supplier No. N04945G
Only null left

Chemical Identifiers

CAS 26830-95-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00056107
InChI Key QGBSLPHQCUIZKK-UHFFFAOYSA-N
Synonym 2-nitro-p-tolunitrile, 2-nitro-4-toluonitrile, benzonitrile, 4-methyl-2-nitro, 2-nitro-4-methylbenzonitrile, 4-methyl-2-nitro-benzonitrile, 4-methyl-2-nitrobenzenecarbonitrile, pubchem13730, acmc-1cjm6, ksc494k5h, 4-methyl-2-nitro benzonitrile
PubChem CID 96235
IUPAC Name 4-methyl-2-nitrobenzonitrile
SMILES CC1=CC=C(C#N)C(=C1)[N+]([O-])=O

Specifications

Melting Point 99°C
Color Yellow
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-p-tolunitrile