missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. N07205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07205G 5 g
N072025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07205G Supplier TCI America Supplier No. N07205G

Chemical Identifiers

CAS 320-94-5
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
InChI Key MYSAXQPTXWKDPQ-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
PubChem CID 642096
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzoic Acid