missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC431740500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431740500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431740500 Supplier Thermo Scientific Chemicals Supplier No. 431740500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21473-01-8 | C10H7BrMg | 231.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21473-01-8 , 109-99-9
Molecular Formula C10H7BrMg
Molecular Weight (g/mol) 231.38
MDL Number MFCD01319910
InChI Key XNUCNPPAQXKAOY-UHFFFAOYSA-M
Synonym 2-naphthylmagnesium bromide, 2-naphthylmagnesiumbromide, 2-naphthyl magnesium bromide, 2-naphthalenylmagnesium bromide, 2-naphthalenyl magnesiumbromide, naphthalen-2-ylmagnesium bromide, xnucnppaqxkaoy-uhfffaoysa-m, bromo naphthalen-2-yl magnesium, 2-naphthylmagnesium bromide, 0.25m in methf, grignard reagent
PubChem CID 11195619
IUPAC Name magnesium;2H-naphthalen-2-ide;bromide
SMILES Br[Mg]C1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 9.76, 85.36
CAS Max % 14.64, 90.24
Color Brown
Density 0.9480g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.948
Solubility Information Solubility in water: reacts.
Formula Weight 231.37
Physical Form Liquid
Chemical Name or Material 2-Naphthylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Susp
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only