missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoyl chloride, 98%

Catalog No. AAA1404630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404630 250 g
AAA1404609 10 g
AAA1404618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404630 Supplier Thermo Scientific Chemicals Supplier No. A1404630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2243-83-6 | C11H7ClO | 190.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2243-83-6
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.626
MDL Number MFCD00004093
InChI Key XNLBCXGRQWUJLU-UHFFFAOYSA-N
Synonym 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
PubChem CID 75246
IUPAC Name naphthalene-2-carbonyl chloride
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)Cl

Specifications

Melting Point 48°C to 52°C
Boiling Point 159°C to 160°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 250 g
UN Number UN3261
Beilstein 907776
Sensitivity Moisture sensitive
Formula Weight 190.63
Percent Purity 98%
Chemical Name or Material 2-Naphthoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-822-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only