missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. N06491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06491G Supplier TCI America Supplier No. N06491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32316-92-0
Molecular Formula C10H9BO2
Molecular Weight (g/mol) 171.99
MDL Number MFCD00236051
InChI Key KPTRDYONBVUWPD-UHFFFAOYSA-N
Synonym 2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol
PubChem CID 2734375
IUPAC Name (naphthalen-2-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 268°C
Color White-Yellow
Quantity 1 g
Formula Weight 171.99
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthaleneboronic Acid (contains varying amounts of Anhydride)