missing translation for 'onlineSavingsMsg'
Learn More

2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine 96.0+%, TCI America™

Catalog No. B18655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18655G 5 g
B186525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18655G Supplier TCI America Supplier No. B18655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 145100-51-2
Molecular Formula C7H3ClF6N2O4S2
Molecular Weight (g/mol) 392.671
MDL Number MFCD00191833
InChI Key TUFGVZMNGTYAQD-UHFFFAOYSA-N
Synonym 2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide
PubChem CID 388544
IUPAC Name N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 392.67
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine [Triflating Reagent]