missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butylfuran, 98%

Catalog No. AAA1337630
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1337630 Supplier Thermo Scientific Chemicals Supplier No. A1337630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4466-24-4 | C8H12O | 124.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4466-24-4
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00047071
InChI Key NWZIYQNUCXUJJJ-UHFFFAOYSA-N
Synonym furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane
PubChem CID 20534
IUPAC Name 2-butylfuran
SMILES CCCCC1=CC=CO1

Specifications

Density 0.886
Boiling Point 153°C to 154°C
Flash Point 32°C (89°F)
Odor Pleasant
Quantity 250 g
UN Number UN1993
Beilstein 107045
Formula Weight 124.18
Percent Purity 98%
Chemical Name or Material 2-n-Butylfuran
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 224-732-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only