missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinopyrimidin-5-yl)methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC69509DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC69509DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

937796-13-9
195.222
KYYQOJQTQOKGPQ-UHFFFAOYSA-N
42556095
C1COCCN1C2=NC=C(C=N2)CO
C9H13N3O2
MFCD09966160
2-morpholinopyrimidin-5-yl methanol, 2-morpholin-4-yl pyrimidin-5-yl methanol, 2-morpholin-4-ylpyrimidin-5-yl methanol, 5-pyrimidinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyrimidin-5-yl methan-1-ol
(2-morpholin-4-ylpyrimidin-5-yl)methanol

Specifications

(2-Morpholinopyrimidin-5-yl)methanol
96.5
97%
C9H13N3O2
MFCD09966160
KYYQOJQTQOKGPQ-UHFFFAOYSA-N
(2-morpholin-4-ylpyrimidin-5-yl)methanol
42556095
97%
937796-13-9
100.0
Amber Glass Bottle
1g
2-morpholinopyrimidin-5-yl methanol, 2-morpholin-4-yl pyrimidin-5-yl methanol, 2-morpholin-4-ylpyrimidin-5-yl methanol, 5-pyrimidinemethanol, 2-4-morpholinyl, 2-morpholin-4-ylpyrimidin-5-yl methan-1-ol
C1COCCN1C2=NC=C(C=N2)CO
195.222
195.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents