missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinobenzylamine, 97%, Thermo Scientific™

Numéro de catalogue. p-2698680
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC24313DA 1g
CC24313DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC24313DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC24313DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 204078-48-8
Formule moléculaire C11H16N2O
Poids moléculaire (g/mol) 192.26
Numéro MDL MFCD03086183
Clé InChI RNYURNUANACIKS-UHFFFAOYSA-N
Synonyme 2-morpholinobenzylamine, 2-morpholinophenyl methanamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-yl benzylamine, 2-4-morpholinyl phenyl methanamine, 1-2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholin-4-ylbenzylamine
PubChem CID 2776563
Nom de l’IUPAC (2-morpholin-4-ylphenyl)methanamine
SOURIRES NCC1=CC=CC=C1N1CCOCC1

Spécifications

Nom ou substance chimique 2-Morpholinobenzylamine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 192.26
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.