missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylvaleraldehyde 95.0+%, TCI America™
$50.33 - $203.17
Chemical Identifiers
| CAS | 123-15-9 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00006986 |
| InChI Key | FTZILAQGHINQQR-UHFFFAOYSA-N |
| Synonym | 2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal |
| PubChem CID | 31245 |
| IUPAC Name | 2-methylpentanal |
| SMILES | CCCC(C)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M059625ML
|
TCI America
M059625ML |
25 mL |
Each for $50.33
|
|
|||||
|
M0596500ML
|
TCI America
M0596500ML |
500 mL |
Each for $203.17
|
|
|||||
Chemical Identifiers
| 123-15-9 | |
| 100.161 | |
| FTZILAQGHINQQR-UHFFFAOYSA-N | |
| 31245 | |
| CCCC(C)C=O |
| C6H12O | |
| MFCD00006986 | |
| 2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal | |
| 2-methylpentanal |
Specifications
| 123-15-9 | |
| 119°C | |
| MFCD00006986 | |
| 2367 | |
| FTZILAQGHINQQR-UHFFFAOYSA-N | |
| 2-methylpentanal | |
| 31245 | |
| ≥95.0% (GC) | |
| 2-Methylvaleraldehyde |
| Colorless | |
| C6H12O | |
| 25 mL | |
| 2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal | |
| CCCC(C)C=O | |
| 100.161 | |
| 100.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : YV4150000
TSCA : Yes