missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylvaleraldehyde 95.0+%, TCI America™
Supplier: TCI America M059625ML
Specifications
| 2-Methylvaleraldehyde | |
| Colorless | |
| C6H12O | |
| 25 mL | |
| 2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal | |
| CCCC(C)C=O | |
| 100.161 | |
| 100.16 | |
| Liquid |
| 123-15-9 | |
| 119°C | |
| MFCD00006986 | |
| 2367 | |
| FTZILAQGHINQQR-UHFFFAOYSA-N | |
| 2-methylpentanal | |
| 31245 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 123-15-9 | |
| 100.161 | |
| FTZILAQGHINQQR-UHFFFAOYSA-N | |
| 31245 | |
| CCCC(C)C=O |
| C6H12O | |
| MFCD00006986 | |
| 2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal | |
| 2-methylpentanal |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : YV4150000
TSCA : Yes