missing translation for 'onlineSavingsMsg'
Learn More

2-Methylvaleraldehyde 95.0+%, TCI America™

Supplier:  TCI America M0596500ML

 View more versions of this product

Catalog No. M0596500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methylvaleraldehyde
Colorless
C6H12O
500 mL
2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal
CCCC(C)C=O
100.161
100.16
Liquid
123-15-9
119°C
MFCD00006986
2367
FTZILAQGHINQQR-UHFFFAOYSA-N
2-methylpentanal
31245
≥95.0% (GC)

Chemical Identifiers

123-15-9
100.161
FTZILAQGHINQQR-UHFFFAOYSA-N
31245
CCCC(C)C=O
C6H12O
MFCD00006986
2-methylvaleraldehyde, methyl valeraldehyde, pentanal, 2-methyl, 2-formylpentane, 2-methylpentaldehyde, valeraldehyde, 2-methyl, 2-methylvaleric aldehyde, pentanal, methyl, alpha-methylpentanal, alpha-methylpentenal
2-methylpentanal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0494

RTECSNumber : YV4150000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions