missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)naphtho[1,2-d]thiazole 98.0+%, TCI America™

Catalog No. M18275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18275G Supplier TCI America Supplier No. M18275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51769-43-8
Molecular Formula C12H9NS2
Molecular Weight (g/mol) 231.331
InChI Key NCJSBVUUUHBCQS-UHFFFAOYSA-N
PubChem CID 103962
IUPAC Name 2-methylsulfanylbenzo[e][1,3]benzothiazole
SMILES CSC1=NC2=C(S1)C=CC3=CC=CC=C32

Specifications

Melting Point 64°C
Color Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 231.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)naphtho[1,2-d]thiazole