missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethylamine, Thermo Scientific™

Numéro de catalogue. CD06838DA
Modifier l'affichage
Click to view available options
Quantité:
1g
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CD06838DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CD06838DA Fournisseur Thermo Scientific Chemicals Code fournisseur CD06838DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 18542-42-2
Formule moléculaire C3H9NS
Poids moléculaire (g/mol) 91.172
Numéro MDL MFCD00014825
Clé InChI CYWGSFFHHMQKET-UHFFFAOYSA-N
Synonyme 2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
PubChem CID 87697
Nom de l’IUPAC 2-methylsulfanylethanamine
SOURIRES CSCCN

Spécifications

Nom ou substance chimique 2-(Methylthio)ethylamine
CAS % minimum 80.0
CAS % maximal 100.0
Densité 0.974g/mL
Fourchette de pourcentages d’essai 95%+
Emballage Amber Glass Bottle
Formule linéaire CH3S(CH2)2NH2
Quantité 1g
Poids de formule 91.18
Pourcentage de pureté Tech.
Niveau Technical
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.